Assay Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261631.1 | NP_001248560.1 | 924 | 91 | ||
| GenBank mRNA | JU320966.1 | - | 920 | 91 | ||
| JV045096.1 | - | 927 | 91 | |||
| JV045097.1 | - | 924 | 91 | |||
More Information
Set Membership: |
|

TOR1AIP2
Genomic Map