Assay Details
| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152338.2 | XP_015007824.2 | 861 | 130 | ||
| GenBank mRNA | JU336413.1 | - | 251 | 130 | ||
| JV723520.1 | - | 250 | 130 | |||
More Information
Set Membership: |
|

BBS10
Genomic Map