Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194274.2 | NP_001181203.1 | 391 | 128 | ||
| GenBank mRNA | JU320863.1 | - | 249 | 128 | ||
| JV661984.1 | - | 189 | 128 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194274.2 transcripts.
Set Membership: |
|

UBTD1
Genomic Map