Assay Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109111.4 | XP_001109111.1 | 1002 | 137 | ||
| XM_015133285.2 | XP_014988771.1 | 1191 | 137 | |||
More Information
Set Membership: |
|

UPK3B
Genomic Map