Assay Details
| Gene Symbol: TEX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145116.2 | XP_015000602.2 | 9728 | 111 | ||
| XM_015145118.2 | XP_015000604.2 | 9888 | 111 | |||
| XM_028852538.1 | XP_028708371.1 | 9702 | 111 | |||
| GenBank mRNA | JV656200.1 | - | 2071 | 111 | ||
More Information
Set Membership: |
|

TEX15
Genomic Map