Assay Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086614.4 | XP_001086614.3 | 1052 | 155 | ||
| XM_015121467.2 | XP_014976953.2 | 1153 | 155 | |||
| XM_015121468.2 | XP_014976954.2 | 1141 | 155 | |||
| XM_015121470.2 | XP_014976956.2 | 1064 | 155 | |||
| XM_015121471.2 | XP_014976957.1 | 946 | 155 | |||
| XM_028837341.1 | XP_028693174.1 | 1109 | 155 | |||
More Information
Set Membership: |
|

SPACA7
Genomic Map