Assay Details
| Gene Symbol: LRTOMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136102.1 | NP_001129574.1 | 82 | 122 | ||
| NM_001199354.1 | NP_001186283.1 | 82 | 122 | |||
| XM_015115045.2 | XP_014970531.1 | 496 | 122 | |||
| XM_015115046.2 | XP_014970532.1 | 513 | 122 | |||
| XM_028832845.1 | XP_028688678.1 | 642 | 122 | |||
| GenBank mRNA | EU627076.1 | - | 82 | 122 | ||
| EU627077.1 | - | 82 | 122 | |||
| EU627078.1 | - | 82 | 122 | |||
| EU627079.1 | - | 82 | 122 | |||
| JV635861.1 | - | 138 | 122 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001136102.1, NM_001199354.1, XM_015115045.2, XM_015115046.2, XM_028832845.1 transcripts.
Set Membership: |
|

LRTOMT
Genomic Map