Assay Details
| Gene Symbol: LRTOMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199354.1 | NP_001186283.1 | 579 | 81 | ||
| XM_015115045.2 | XP_014970531.1 | 993 | 81 | |||
| XM_015115046.2 | XP_014970532.1 | 1010 | 81 | |||
| XM_015115047.2 | XP_014970533.1 | 588 | 81 | |||
| XM_015115048.2 | XP_014970534.1 | 810 | 81 | |||
| XM_015115049.2 | XP_014970535.1 | 577 | 81 | |||
| XM_015115050.2 | XP_014970536.1 | 995 | 81 | |||
| GenBank mRNA | EU627076.1 | - | 579 | 81 | ||
| JV046448.1 | - | 488 | 81 | |||
| JV635861.1 | - | 635 | 81 | |||
More Information
Set Membership: |
|

LRTOMT
Genomic Map