Assay Details
| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094766.4 | XP_001094766.2 | 512 | 133 | ||
| GenBank mRNA | JV707571.1 | - | 329 | 133 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF597| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094766.4 | XP_001094766.2 | 512 | 133 | ||
| GenBank mRNA | JV707571.1 | - | 329 | 133 | ||
Set Membership: |
|