Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261504.1 | NP_001248433.1 | 86 | 130 | ||
| GenBank mRNA | JU322920.1 | - | 86 | 130 | ||
| JV510331.1 | - | 90 | 130 | |||
More Information
Set Membership: |
|

SIAE
Genomic Map