Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113056.4 | XP_001113056.3 | 2610 | 103 | ||
| XM_015134461.2 | XP_014989947.2 | 2615 | 103 | |||
| XM_015134462.2 | XP_014989948.2 | 3028 | 103 | |||
| XM_028845589.1 | XP_028701422.1 | 2906 | 103 | |||
| XM_028845590.1 | XP_028701423.1 | 2328 | 103 | |||
| XM_028845591.1 | XP_028701424.1 | 3022 | 103 | |||
| GenBank mRNA | JU324067.1 | - | 2322 | 103 | ||
More Information
Set Membership: |
|

NYAP1
Genomic Map