Assay Details
| Gene Symbol: NEGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261637.1 | NP_001248566.1 | 497 | 106 | ||
| XM_015142298.2 | XP_014997784.2 | 3746 | 106 | |||
| GenBank mRNA | JU321197.1 | - | 497 | 106 | ||
| JV611985.1 | - | 512 | 106 | |||
More Information
Set Membership: |
|

NEGR1
Genomic Map