Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261100.1 | NP_001248029.1 | 967 | 141 | ||
| XM_015112508.2 | XP_014967994.1 | 829 | 141 | |||
| GenBank mRNA | HM370982.1 | - | 478 | 141 | ||
| JU320424.1 | - | 934 | 141 | |||
| JU320425.1 | - | 934 | 141 | |||
| JU470584.1 | - | 941 | 141 | |||
| JU470585.1 | - | 941 | 141 | |||
| JV044072.1 | - | 967 | 141 | |||
| JV044073.1 | - | 967 | 141 | |||
| JV566547.1 | - | 1128 | 141 | |||
More Information
Set Membership: |
|

SPRED2
Genomic Map