Assay Details
| Gene Symbol: CCDC144NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836122.1 | XP_028691955.1 | 3427 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC144NL| Gene Symbol: CCDC144NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836122.1 | XP_028691955.1 | 3427 | 78 | ||
Set Membership: |
|