Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260842.1 | NP_001247771.1 | 710 | 62 | ||
| XM_015139851.2 | XP_014995337.1 | 770 | 62 | |||
| XM_028840504.1 | XP_028696337.1 | 746 | 62 | |||
| XM_028840505.1 | XP_028696338.1 | 754 | 62 | |||
| GenBank mRNA | JU332890.1 | - | 727 | 62 | ||
| JU473419.1 | - | 734 | 62 | |||
| JV044791.1 | - | 710 | 62 | |||
More Information
Set Membership: |
|

TMEM219
Genomic Map