Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194382.2 | NP_001181311.2 | 247 | 143 | ||
| XM_028835691.1 | XP_028691524.1 | 286 | 143 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM95
Genomic Map