Assay Details
| Gene Symbol: KRT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119637.2 | XP_014975123.1 | 917 | 79 | ||
| GenBank mRNA | JU486244.1 | - | 416 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT9| Gene Symbol: KRT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119637.2 | XP_014975123.1 | 917 | 79 | ||
| GenBank mRNA | JU486244.1 | - | 416 | 79 | ||
Set Membership: |
|