Assay Details
Gene Symbol: SLC26A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141224.2 | XP_014996710.1 | 237 | 152 | ||
XM_028849833.1 | XP_028705666.1 | 237 | 152 | |||
GenBank mRNA | JV053782.1 | - | 143 | 152 | ||
JV644329.1 | - | 114 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015141224.2, XM_028849833.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
