Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849252.1 | XP_028705085.1 | 787 | 125 | ||
| XM_028849253.1 | XP_028705086.1 | 893 | 125 | |||
| XM_028849254.1 | XP_028705087.1 | 623 | 125 | |||
| XM_028849255.1 | XP_028705088.1 | 643 | 125 | |||
| XM_028849256.1 | XP_028705089.1 | 550 | 125 | |||
| XM_028849257.1 | XP_028705090.1 | 774 | 125 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028849252.1, XM_028849253.1, XM_028849254.1, XM_028849255.1, XM_028849256.1, XM_028849257.1 transcripts.
Set Membership: |
|

ANKRD34B
Genomic Map