Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040278.2 | NP_001035368.1 | 193 | 90 | ||
| XM_015123564.2 | XP_014979050.1 | 210 | ||||
| GenBank mRNA | AY585862.1 | - | 146 | 90 | ||
| JU323488.1 | - | 187 | 90 | |||
| JV045060.1 | - | 193 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COX7A1
Genomic Map