Assay Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266832.1 | NP_001253761.1 | 135 | 87 | ||
| GenBank mRNA | JU321390.1 | - | 121 | 87 | ||
| JU473198.1 | - | 135 | 87 | |||
More Information
Set Membership: |
|

TCAP
Genomic Map