Assay Details
| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093356.4 | XP_001093356.3 | 935 | 107 | ||
| GenBank mRNA | JU328639.1 | - | 77 | 107 | ||
| JV600087.1 | - | 259 | 107 | |||
| JV635290.1 | - | 510 | 107 | |||
More Information
Set Membership: |
|

HS3ST2
Genomic Map