Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089248.4 | XP_001089248.2 | 326 | 148 | ||
| XM_015152360.2 | XP_015007846.1 | 326 | 148 | |||
| GenBank mRNA | JU323485.1 | - | 163 | 148 | ||
| JU485101.1 | - | 74 | 148 | |||
More Information
Set Membership: |
|

CCDC59
Genomic Map