Assay Details
| Gene Symbol: GPRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147053.2 | XP_015002539.2 | 215 | 140 | ||
| XM_015147054.2 | XP_015002540.2 | 1194 | 140 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015147053.2, XM_015147054.2 transcripts.
Set Membership: |
|

GPRIN2
Genomic Map