Assay Details
Gene Symbol: JPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039945.2 | NP_001035034.1 | 359 | 106 | ||
GenBank mRNA | CO583448.1 | - | 383 | 106 | ||
JU320949.1 | - | 407 | 106 | |||
JU478323.1 | - | 344 | 106 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
