Assay Details
| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265823.1 | NP_001252752.1 | 1085 | 164 | ||
| GenBank mRNA | JV600257.1 | - | 1085 | 164 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMX1| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265823.1 | NP_001252752.1 | 1085 | 164 | ||
| GenBank mRNA | JV600257.1 | - | 1085 | 164 | ||
Set Membership: |
|