Assay Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126723.2 | XP_014982209.2 | 289 | 138 | ||
XM_015126724.2 | XP_014982210.2 | 289 | 138 | |||
GenBank mRNA | JU321025.1 | - | 255 | 138 | ||
JU472237.1 | - | 252 | 138 | |||
JV045023.1 | - | 251 | 138 | |||
JV566929.1 | - | 253 | 138 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
