Assay Details
| Gene Symbol: CLDN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134531.2 | XP_014990017.1 | 2039 | 93 | ||
| XM_015134532.2 | XP_014990018.1 | 1999 | 93 | |||
| GenBank mRNA | JU333763.1 | - | 277 | 93 | ||
| JU471456.1 | - | 1045 | 93 | |||
More Information
Set Membership: |
|

CLDN15
Genomic Map