Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260770.1 | NP_001247699.1 | 365 | 108 | ||
| XM_015145630.2 | XP_015001116.1 | 852 | 108 | |||
| XM_015145631.2 | XP_015001117.1 | 538 | 108 | |||
| XM_015145632.2 | XP_015001118.1 | 390 | 108 | |||
| GenBank mRNA | JU336215.1 | - | 372 | 108 | ||
| JU336216.1 | - | 372 | 108 | |||
| JU477057.1 | - | 349 | 108 | |||
| JV048147.1 | - | 365 | 108 | |||
| JV660467.1 | - | 553 | 108 | |||
More Information
Set Membership: |
|

OSGIN2
Genomic Map