Assay Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260582.1 | NP_001247511.1 | 1823 | 118 | ||
| XM_028841751.1 | XP_028697584.1 | 1673 | 118 | |||
| GenBank mRNA | JU335484.1 | - | 1831 | 118 | ||
| JU335487.1 | - | 1831 | 118 | |||
| JU476433.1 | - | 1742 | 118 | |||
| JU476434.1 | - | 1742 | 118 | |||
| JV047676.1 | - | 1823 | 118 | |||
| JV047677.1 | - | 1823 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRPS1
Genomic Map