Assay Details
| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032915.1 | NP_001028087.1 | 188 | 125 | ||
| GenBank mRNA | AY656809.2 | - | 188 | 125 | ||
| JV667310.1 | - | 148 | 125 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MIF
Genomic Map