Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040238.1 | NP_001035328.1 | 831 | 76 | ||
| XM_015142702.2 | XP_014998188.2 | 971 | 76 | |||
| XM_015142703.2 | XP_014998189.2 | 969 | 76 | |||
| GenBank mRNA | AY635458.1 | - | 831 | 76 | ||
More Information
Set Membership: |
|

CYP1A1
Genomic Map