Assay Details
| Gene Symbol: HES5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136275.2 | XP_014991761.2 | 1099 | 64 | ||
| XM_015136334.2 | XP_014991820.1 | 748 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HES5| Gene Symbol: HES5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136275.2 | XP_014991761.2 | 1099 | 64 | ||
| XM_015136334.2 | XP_014991820.1 | 748 | 64 | |||
Set Membership: |
|