Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194260.2 | NP_001181189.2 | 549 | 69 | ||
| GenBank mRNA | JU323086.1 | - | 527 | 69 | ||
| JU472356.1 | - | 547 | 69 | |||
| JV044947.1 | - | 522 | 69 | |||
More Information
Set Membership: |
|

MYG1
Genomic Map