Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194454.1 | NP_001181383.1 | 63 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194454.1 transcripts.
Set Membership: |
|

FAM83B
Genomic Map