Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194454.1 | NP_001181383.1 | 574 | 112 | ||
| XM_028846786.1 | XP_028702619.1 | 567 | 112 | |||
More Information
Set Membership: |
|

FAM83B
Genomic Map