Assay Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190916.1 | NP_001177845.1 | 189 | 66 | ||
| XM_015149072.2 | XP_015004558.1 | 278 | 66 | |||
| GenBank mRNA | CO774585.1 | - | 193 | 66 | ||
| JU320945.1 | - | 263 | 66 | |||
| JU320946.1 | - | 258 | 66 | |||
More Information
Set Membership: |
|

ROMO1
Genomic Map