Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193364.1 | NP_001180293.1 | 62 | 102 | ||
| GenBank mRNA | CD766914.1 | - | 66 | 102 | ||
| JU332262.1 | - | 125 | 102 | |||
| JU332263.1 | - | 125 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193364.1 transcripts.
Set Membership: |
|

FAM229B
Genomic Map