Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193364.1 | NP_001180293.1 | 219 | 98 | ||
| XM_015137241.2 | XP_014992727.1 | 261 | 98 | |||
| XM_015137242.2 | XP_014992728.1 | 252 | 98 | |||
| XM_015137243.2 | XP_014992729.1 | 257 | 98 | |||
| XM_015137244.2 | XP_014992730.1 | 170 | 98 | |||
| XM_015137245.1 | XP_014992731.1 | 244 | 98 | |||
| XM_015137246.2 | XP_014992732.1 | 170 | 98 | |||
| XM_015137247.2 | XP_014992733.1 | 170 | 98 | |||
| XM_015137248.2 | XP_014992734.1 | 170 | 98 | |||
| XM_028846600.1 | XP_028702433.1 | 229 | 98 | |||
| XM_028846601.1 | XP_028702434.1 | 169 | 98 | |||
| GenBank mRNA | CD766914.1 | - | 223 | 98 | ||
| JU332262.1 | - | 282 | 98 | |||
| JU332263.1 | - | 282 | 98 | |||
| JV044710.1 | - | 168 | 98 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193364.1, XM_015137241.2, XM_015137242.2, XM_015137243.2, XM_015137244.2, XM_015137245.1, XM_015137246.2, XM_015137247.2, XM_015137248.2, XM_028846600.1, XM_028846601.1 transcripts.
Set Membership: |
|

FAM229B
Genomic Map