Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193364.1 | NP_001180293.1 | 363 | 83 | ||
| XM_015137241.2 | XP_014992727.1 | 405 | 83 | |||
| XM_015137242.2 | XP_014992728.1 | 396 | 83 | |||
| XM_015137243.2 | XP_014992729.1 | 401 | 83 | |||
| XM_015137244.2 | XP_014992730.1 | 314 | 83 | |||
| XM_015137245.1 | XP_014992731.1 | 388 | 83 | |||
| XM_015137246.2 | XP_014992732.1 | 314 | 83 | |||
| XM_015137247.2 | XP_014992733.1 | 314 | 83 | |||
| XM_015137248.2 | XP_014992734.1 | 314 | 83 | |||
| XM_028846600.1 | XP_028702433.1 | 373 | 83 | |||
| XM_028846601.1 | XP_028702434.1 | 313 | 83 | |||
| XM_028846602.1 | XP_028702435.1 | 274 | 83 | |||
| GenBank mRNA | CD766914.1 | - | 367 | 83 | ||
| JU332262.1 | - | 426 | 83 | |||
| JU332263.1 | - | 426 | 83 | |||
| JV044710.1 | - | 312 | 83 | |||
More Information
Set Membership: |
|

FAM229B
Genomic Map