Assay Details
| Gene Symbol: MRPL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193734.2 | NP_001180663.1 | 343 | 84 | ||
| GenBank mRNA | JU335862.1 | - | 272 | 84 | ||
| JV047891.1 | - | 259 | 84 | |||
| JV601176.1 | - | 292 | 84 | |||
More Information
Set Membership: |
|

MRPL17
Genomic Map