Assay Details
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194764.2 | NP_001181693.1 | 207 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194764.2 transcripts.
Set Membership: |
![]() ![]() |
