Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194373.2 | NP_001181302.1 | 953 | 99 | ||
| GenBank mRNA | JU472795.1 | - | 930 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRP7A| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194373.2 | NP_001181302.1 | 953 | 99 | ||
| GenBank mRNA | JU472795.1 | - | 930 | 99 | ||
Set Membership: |
|