Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257304.2 | NP_001244233.2 | 847 | 57 | ||
| GenBank mRNA | HQ423394.1 | - | 839 | 57 | ||
| HQ423395.1 | - | 836 | 57 | |||
| HQ423396.1 | - | 836 | 57 | |||
| JQ037994.2 | - | 847 | 57 | |||
| JQ037995.1 | - | 847 | 57 | |||
| JQ037996.2 | - | 847 | 57 | |||
| JU333388.1 | - | 881 | 57 | |||
| JV489157.1 | - | 853 | 57 | |||
More Information
Set Membership: |
|

FCGR1A
Genomic Map