Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260589.1 | NP_001247518.1 | 325 | 113 | ||
| XM_028835761.1 | XP_028691594.1 | 630 | 113 | |||
| XM_028835762.1 | XP_028691595.1 | 471 | 113 | |||
| XM_028835763.1 | XP_028691596.1 | 330 | 113 | |||
| XM_028835764.1 | XP_028691597.1 | 1656 | 113 | |||
| GenBank mRNA | JU336515.1 | - | 350 | 113 | ||
| JU477326.1 | - | 325 | 113 | |||
| JV629726.1 | - | 277 | 113 | |||
More Information
Set Membership: |
|

LIMD2
Genomic Map