Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261166.1 | NP_001248095.1 | 1022 | 93 | ||
| XM_028849252.1 | XP_028705085.1 | 1816 | 93 | |||
| XM_028849253.1 | XP_028705086.1 | 1922 | 93 | |||
| XM_028849254.1 | XP_028705087.1 | 1652 | 93 | |||
| XM_028849255.1 | XP_028705088.1 | 1672 | 93 | |||
| XM_028849256.1 | XP_028705089.1 | 1579 | 93 | |||
| XM_028849257.1 | XP_028705090.1 | 1803 | 93 | |||
| GenBank mRNA | JU322876.1 | - | 1022 | 93 | ||
More Information
Set Membership: |
|

ANKRD34B
Genomic Map