Assay Details
| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261357.2 | NP_001248286.1 | 2326 | 89 | ||
| XM_015143571.2 | XP_014999057.1 | 2642 | 89 | |||
| XM_015143580.2 | XP_014999066.1 | 3001 | 89 | |||
| XM_015143587.2 | XP_014999073.1 | 2778 | 89 | |||
| XM_015143593.2 | XP_014999079.1 | 2511 | 89 | |||
| XM_028849267.1 | XP_028705100.1 | 5400 | 89 | |||
| XM_028849290.1 | XP_028705123.1 | 2568 | 89 | |||
| GenBank mRNA | JU332225.1 | - | 2327 | 89 | ||
| JU332226.1 | - | 2327 | 89 | |||
| JU332227.1 | - | 2327 | 89 | |||
| JU332228.1 | - | 2327 | 89 | |||
| JU333712.1 | - | 2327 | 89 | |||
| JU333713.1 | - | 2327 | 89 | |||
| JU333714.1 | - | 2327 | 89 | |||
| JU333715.1 | - | 2327 | 89 | |||
| JU333716.1 | - | 2327 | 89 | |||
More Information
Set Membership: |
|

LRRC8B
Genomic Map