Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261602.1 | NP_001248531.1 | 373 | 94 | ||
| XM_028852130.1 | XP_028707963.1 | 457 | 94 | |||
| XM_028852131.1 | XP_028707964.1 | 387 | 94 | |||
| GenBank mRNA | JU332985.1 | - | 351 | 94 | ||
| JU332986.1 | - | 351 | 94 | |||
| JU332987.1 | - | 351 | 94 | |||
| JU332988.1 | - | 351 | 94 | |||
| JV046225.1 | - | 373 | 94 | |||
| JV642643.1 | - | 381 | 94 | |||
More Information
Set Membership: |
|

UBXN2B
Genomic Map