Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261685.2 | NP_001248614.1 | 1222 | 69 | ||
| XM_015116859.2 | XP_014972345.1 | 1336 | 69 | |||
| XM_015116860.2 | XP_014972346.1 | 1375 | 69 | |||
| GenBank mRNA | JU320681.1 | - | 1286 | 69 | ||
| JU472142.1 | - | 1278 | 69 | |||
| JV045300.1 | - | 1223 | 69 | |||
More Information
Set Membership: |
|

ZBTB26
Genomic Map