Assay Details
| Gene Symbol: CALHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265885.1 | NP_001252814.1 | 810 | 97 | ||
| XM_015148020.2 | XP_015003506.1 | 1937 | 97 | |||
| XM_015148021.2 | XP_015003507.1 | 1237 | 97 | |||
| XM_015148022.2 | XP_015003508.1 | 1117 | 97 | |||
| XM_015148023.2 | XP_015003509.1 | 1817 | 97 | |||
| XM_028853521.1 | XP_028709354.1 | 1218 | 97 | |||
| XM_028853522.1 | XP_028709355.1 | 1918 | 97 | |||
| GenBank mRNA | JV508674.1 | - | 580 | 97 | ||
| JV635793.1 | - | 810 | 97 | |||
More Information
Set Membership: |
|

CALHM2
Genomic Map